Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-2,6-dichloroisonicotinate
tert-Butyl-2,6-dichloroisonicotinate 97%
tert-Butyl-2,6-dichloroisonicotinate 97%
APOSOR6618-5G
:
Apollo Scientific
:
tert-Butyl-2,6-dichloroisonicotinate 97%
APOSOR6618-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
248.10 g/mol
:
Ambient
:
75308-46-2
:
C₁₀H₁₁Cl₂NO₂
:
MFCD08445655