3-Bromo-4-methylpyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3430-22-6
:
Formula:C₆H₆BrN
MDL Number:MFCD00082592
:172.02 g/mol
Boiling Pt:199…200 °C
Density:1.549 g/cm³ (25 °C)
Flash Pt:79 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together