Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nitro-m-toluic acid
3-Methyl-2-nitrobenzoic acid
3-Methyl-2-nitrobenzoic acid
3-Methyl-2-nitrobenzoic acid
APOSOR61446-25G
:
Apollo Scientific
:
3-Methyl-2-nitrobenzoic acid
APOSOR61446-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
181.15 g/mol
:
220…223 °C
:
226-610-9
:
GHS07
:
Ambient
:
5437-38-7
:
C₈H₇NO₄
:
MFCD00007180