Order Entry
France
ContactUsLinkComponent
(S)-4-Hydroxypyrrolidine-2-one
Irritant
(S)-4-Hydroxypyrrolidine-2-one
Catalog # APOSOR60184-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-4-Hydroxypyrrolidine-2-one
Catalog # APOSOR60184-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :101.105 g/mol
Melting Pt:156…159 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:68108-18-9
Formula:C₄H₇NO₂
MDL Number:MFCD00273370