Piperazine-1-carboximidamide
Catalog # APOSOR60177-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:128.18 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:45695-84-9
Formula:C₅H₁₂N₄
MDL Number:MFCD04116543