
(6,7-Dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-3-yl)methanamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:923156-44-9
:
Formula:C₆H₁₀N₄
MDL Number:MFCD08691190
:138.170 g/mol
Melting Pt:79 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together