Fumaric acid monomethyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2756-87-8
:
Formula:C₅H₆O₄
MDL Number:MFCD00063174
:130.1 g/mol
Melting Pt:143…147 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together