Fumaric acid monoethyl ester
Catalog # APOSOR59886-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:144.13 g/mol
Melting Pt:64…67 °C
Storage Temperature:Ambient
CAS Number:2459-05-4
Formula:C₆H₈O₄
MDL Number:MFCD00002699