
3,4-Dimethoxyphenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:122775-35-3
:
Formula:C₈H₁₁BO₄
MDL Number:MFCD01074574
:181.98 g/mol
Melting Pt:245…250 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together