
Bis(pinacolato)diboron 99%
:73183-34-3
:
:C₁₂H₂₄B₂O₄
:MFCD00799570
:137…140 °C
:253.94 g/mol
:GHS07
:Refrigerator
Frequently Bought Together
2-Propanol ≥99.5% ACS, Riedel-de Haën™
: Honeywell Chemicals
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
: PanReac AppliChem
Triethylsilane 98%
: Apollo Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99.5% for HPLC
: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
4-Methyl-2-pentanone ≥97%, TECHNICAL
: VWR Chemicals
Dichloromethane 99.8% stabilised for HPLC
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Toluene ≥99.7% for spectrophotometry
: Thermo Fisher Scientific