Order Entry
France
ContactUsLinkComponent
4-Nitroaniline 98%
Toxic
Health
4-Nitroaniline 98%
Catalog # APOSOR5450-25G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Health
4-Nitroaniline 98%
Catalog # APOSOR5450-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :138.13 g/mol
Boiling Pt:332 °C
Melting Pt:147…149 °C
Density:1.44 g/cm³ (25 °C)
Flash Pt:165 °C
Storage Temperature:Ambient
EINECS:20-281-0
UN:1661
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:100-01-6
Formula:C₆H₆N₂O₂
MDL Number:MFCD00007858