5-Methylpyridine-2-sulphonamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65938-77-4
:
Formula:C₆H₈N₂O₂S
MDL Number:MFCD00661363
:172.21 g/mol
Melting Pt:127…132 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together