Order Entry
France
ContactUsLinkComponent
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
Catalog # APOSOR51749-25G
Supplier:  Apollo Scientific
CAS Number:  
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
Catalog # APOSOR51749-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :217.27 g/mol
Melting Pt:81…86 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:61478-26-0
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD02094386