Order Entry
France
ContactUsLinkComponent
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Catalog # APOSOR480762-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-1-boc-3-pyrrolidinecarboxylate 97%
Catalog # APOSOR480762-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :229.28 g/mol
Boiling Pt:105 °C/0.2mmHg
Density:1.08 g/cm³ (25 °C)
Storage Temperature:Refrigerator
CAS Number:122684-33-7
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD04038683