Order Entry
France
ContactUsLinkComponent
3,5-Dibromothiophene-2-carboxylic acid 95%
3,5-Dibromothiophene-2-carboxylic acid 95%
Catalog # APOSOR480644-5G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dibromothiophene-2-carboxylic acid 95%
Catalog # APOSOR480644-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :285.95 g/mol
Storage Temperature:Ambient
CAS Number:7311-68-4
Formula:C₅H₂Br₂O₂S
MDL Number:MFCD01006744