
Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%
Catalog # APOSOR480237-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:144.13 g/mol
Boiling Pt:85…87 °C
Melting Pt:47…51 °C
:GHS07
Storage Temperature:Ambient
CAS Number:113020-21-6
Formula:C₆H₈O₄
MDL Number:MFCD06800496