N-Boc-4-oxo-L-proline-tert-butyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:166410-05-5
:
Formula:C₁₄H₂₃NO₅
MDL Number:MFCD06797013
:285.35 g/mol
Melting Pt:67…68 °C
Storage Temperature:Refrigerator
Frequently Bought Together