Order Entry
France
ContactUsLinkComponent
(Chloromethyl)cyclopropane 95%
Flammable
Irritant
(Chloromethyl)cyclopropane 95%
Catalog # APOSOR460074-25G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
(Chloromethyl)cyclopropane 95%
Catalog # APOSOR460074-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :90.55 g/mol
Boiling Pt:87…89 °C
Density:0.98 g/cm³ (25 °C)
Flash Pt:–2 °C
UN:1993
ADR:3,II
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:5911-08-0
Formula:C₄H₇Cl
MDL Number:MFCD00001307