Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromoisatoic anhydride
5-Bromoisatoic anhydride 95+%
5-Bromoisatoic anhydride 95+%
5-Bromoisatoic anhydride 95+%
APOSOR4597-5G
:
Apollo Scientific
:
5-Bromoisatoic anhydride 95+%
APOSOR4597-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
242.03 g/mol
:
285…290 °C (dec.)
:
GHS07
:
GHS08
:
Ambient
:
4692-98-2
:
C₈H₄BrNO₃
:
MFCD00016921