Diethyl fumarate 95%
Catalog # APOSOR4507-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:172.18 g/mol
Boiling Pt:218…219 °C
Melting Pt:44228 °C
Density:1.052 g/cm³ (25 °C)
Flash Pt:92 °C
Storage Temperature:Ambient
EINECS:210-819-7
:GHS07
CAS Number:623-91-6
Formula:C₈H₁₂O₄
MDL Number:MFCD00064455