Order Entry
France
Orders LinkContactUsLinkComponent
ε-Caprolactam 99%

ε-Caprolactam 99%

Irritant
Supplier:   Apollo Scientific
ε-Caprolactam 99%
Supplier:  Apollo Scientific
Certificates
ε-Caprolactam 99%

4 Options Available Below

Product Details & Documents
Synonyms:  Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam
  • CAS Number:105-60-2
  • Formula:C₆H₁₁NO
  • MDL Number:MFCD00006936
  • Molecular Weight:113.16 g/mol
  • Boiling Pt:267 °C
  • Melting Pt:70 °C
  • Flash Pt:130 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 21,80 €
    21,80 €
    1 * 100 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 57,90 €
    57,90 €
    1 * 500 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 261,00 €
    261,00 €
    1 * 2,5 kg
       261,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 2 540,00 €
    2 540,00 €
    1 * 25 kg
       2 540,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.