5-Methylpyridin-3-ol
Catalog # APOSOR40438-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Melting Pt:136…140 °C
:109.13 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:42732-49-0
Formula:C₆H₇NO
MDL Number:MFCD00661297