Order Entry
France
ContactUsLinkComponent
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%
Catalog # APOSOR350462-1G
Supplier:  Apollo Scientific
CAS Number:  
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%
Catalog # APOSOR350462-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.66 g/mol
Storage Temperature:Ambient
CAS Number:19017-52-8
Formula:C₁₁H₁₀ClNO₂
MDL Number:MFCD00152035