N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboxamidine
Catalog # APOSOR350307-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:210.24 g/mol
Storage Temperature:Ambient
CAS Number:152120-61-1
Formula:C₉H₁₄N₄O₂
MDL Number:MFCD00216663