Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
APOSOR350259-500G
:
Apollo Scientific
:
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
APOSOR350259-500G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 g
Seq:
3645
:
:
143.14 g/mol
:
90 °C/ 0.1mmHg
:
1.226 g/cm³ (25 °C)
:
113 °C
:
225-567-3
:
GHS07
:
Ambient
:
4931-66-2
:
C₆H₉NO₃
:
MFCD00080931