Order Entry
France
ContactUsLinkComponent
2-Acetyl-4-bromopyridine 95+%
2-Acetyl-4-bromopyridine 95+%
Catalog # APOSOR346559-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Acetyl-4-bromopyridine 95+%
Catalog # APOSOR346559-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :200.035 g/mol
Storage Temperature:Ambient
CAS Number:1060805-69-7
Formula:C₇H₆BrNO
MDL Number:MFCD13189212