Order Entry
France
ContactUsLinkComponent
Methyl-6-(hydroxymethyl)nicotinate 95%
Methyl-6-(hydroxymethyl)nicotinate 95%
Catalog # APOSOR346326-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-6-(hydroxymethyl)nicotinate 95%
Catalog # APOSOR346326-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.1600 g/mol
Melting Pt:77 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:56026-36-9
Formula:C₈H₉NO₃
MDL Number:MFCD00643821