N'-[1-Amino-2-(4-methoxyphenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053655-77-8
:
Formula:C₁₄H₂₁N₃O₄
MDL Number:MFCD10568183
:295.34 g/mol
Storage Temperature:Ambient
Frequently Bought Together