Order Entry
France
ContactUsLinkComponent
 
6-Hydroxy-3-iodo-2-methylpyridine 98%
Catalog # APOSOR345454-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Hydroxy-3-iodo-2-methylpyridine 98%
Catalog # APOSOR345454-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.02 g/mol
Storage Temperature:Ambient
CAS Number:927870-76-6
Formula:C₆H₆INO
MDL Number:MFCD09037468