Order Entry
France
ContactUsLinkComponent
2-Amino-6-methylpyridin-3-ol 97%
2-Amino-6-methylpyridin-3-ol 97%
Catalog # APOSOR345045-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-6-methylpyridin-3-ol 97%
Catalog # APOSOR345045-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :124.1400 g/mol
Storage Temperature:Ambient
CAS Number:20348-16-7
Formula:C₆H₈N₂O
MDL Number:MFCD09839245