Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3-cyanopyrazine
2-Chloro-3-cyanopyrazine 98%
2-Chloro-3-cyanopyrazine 98%
APOSOR3400-5G
:
Apollo Scientific
:
2-Chloro-3-cyanopyrazine 98%
APOSOR3400-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
139.5400 g/mol
:
38…40 °C
:
Ambient
:
3439
:
6.1,III
:
GHS07
:
55557-52-3
:
C₅H₂ClN₃
:
MFCD00219653