Order Entry
France
ContactUsLinkComponent
(5-Methylpyridin-2-yl)methanol 95%
(5-Methylpyridin-2-yl)methanol 95%
Catalog # APOSOR33603-1G
Supplier:  Apollo Scientific
CAS Number:  
(5-Methylpyridin-2-yl)methanol 95%
Catalog # APOSOR33603-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :123.16 g/mol
Storage Temperature:Ambient
CAS Number:22940-71-2
Formula:C₇H₉NO
MDL Number:MFCD09999992