5-Methoxy-1-(2-pyrimidinyl)-1H-indole-3-carbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886361-96-2
:
Formula:C₁₄H₁₁N₃O₂
MDL Number:MFCD08056657
:253.26 g/mol
Storage Temperature:Ambient
Frequently Bought Together