Pyrrolidine-1-carboxamidinium acetate
Catalog # APOSOR3256-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.22 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:91983-81-2
Formula:C₇H₁₅N₃O₂
MDL Number:MFCD09702434