N'-Cyano-4-pyridinecarboximidamide
Catalog # APOSOR3251-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:23275-43-6
Formula:C₇H₆N₄
MDL Number:MFCD11553048