7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester
Catalog # APOSOR317185-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:226.28 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1234616-51-3
Formula:C₁₁H₁₈N₂O₃
MDL Number:MFCD18325190