Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2-methylpyridine
3-Chloro-2-methylpyridine
3-Chloro-2-methylpyridine
APOSOR315077-1G
:
Apollo Scientific
:
3-Chloro-2-methylpyridine
APOSOR315077-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
127.5700 g/mol
:
Ambient
:
72093-03-9
:
C6H6ClN
:
MFCD04114121