Benzyl isothiocyanate
Bottle, 1 g
Catalog # APOSOR3098-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Benzylisothiocyanate
- CAS Number:622-78-6
- Formula:C₈H₇NS
- MDL Number:MFCD00004819
- Molecular Weight:149.22 g/mol
- Boiling Pt:242…243 °C
- Density:1.125 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- UN:3082
- ADR:9+EHS,III
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



