2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-tert-butyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1211526-53-2
:
Formula:C₁₂H₁₉NO₄
MDL Number:MFCD17016194
:241.29 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together