
5-Chloro-2-methoxy-nicotinic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54916-65-3
:
Formula:C₇H₆ClNO₃
MDL Number:MFCD08236790
:187.5804 g/mol
Melting Pt:146…148 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together