Order Entry
France
ContactUsLinkComponent
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Catalog # APOSOR307618-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Catalog # APOSOR307618-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:30740-95-5
Formula:C₁₂H₁₆ClNO₄
MDL Number:MFCD06661901