2-Amino-5-methylpyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1603-41-4
:
Formula:C₆H₈N₂
MDL Number:MFCD00006328
:108.14 g/mol
Boiling Pt:226…228 °C
Melting Pt:74…78 °C
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:216-503-5
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together