Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-tert-Butyl-2,6-dimethylaniline
4-tert-Butyl-2,6-dimethylaniline 98%
4-tert-Butyl-2,6-dimethylaniline 98%
APOSOR27022-5G
:
Apollo Scientific
:
4-tert-Butyl-2,6-dimethylaniline 98%
APOSOR27022-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
96 °C/1mmHg
:
Ambient
:
2810
:
6.1,III
:
177.29 g/mol
:
GHS07
:
42014-60-8
:
C₁₂H₁₉N
:
MFCD00052181