Furan-3-carboximidamide hydrochloride, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54649-21-7
:
Formula:C₅H₇ClN₂O
MDL Number:MFCD03086209
:146.58 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together