Order Entry
France
ContactUsLinkComponent
 
 
Diethyl cyclobutane-1,1-dicarboxylate 98%
  APOSOR24547-25G
 :  
 
Diethyl cyclobutane-1,1-dicarboxylate 98%
  APOSOR24547-25G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

 :
 :226 °C
 :1.05 g/cm³ (25 °C)
 :89 °C
 :200.24 g/mol
 :223-239-4
 :GHS07
 :Ambient
 :3779-29-1
 :C₁₀H₁₆O₄
 :MFCD00019261