
4,5-Dibromo-2-thiophenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:81606-31-7
:
Formula:C₄HBr₂ClO₂S₂
MDL Number:MFCD00041439
:340.44 g/mol
Melting Pt:75…79 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together