Order Entry
France
Orders LinkContactUsLinkComponent
Chroman-4-one

Chroman-4-one

Bottle, 25 g
Catalog # APOSOR22417-25G
Irritant
Supplier:   Apollo Scientific
Chroman-4-one
Catalog # APOSOR22417-25G
Supplier:  Apollo Scientific
CAS Number:  491-37-2
Chroman-4-one

Bottle, 25 g

CAS Number:  491-37-2
Price
50,70 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Chromanone
  • CAS Number:491-37-2
  • Formula:C₉H₈O₂
  • MDL Number:MFCD00006840
  • Molecular Weight:148.16 g/mol
  • Boiling Pt:127…128 °C/13mmHg
  • Melting Pt:35…38 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:207-736-3
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.