Order Entry
France
ContactUsLinkComponent
6-Chloro-3-pyridineacetic acid 95%
Irritant
6-Chloro-3-pyridineacetic acid 95%
Catalog # APOSOR2190-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Chloro-3-pyridineacetic acid 95%
Catalog # APOSOR2190-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :171.58 g/mol
Melting Pt:169…176 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:39891-13-9
Formula:C₇H₆ClNO₂
MDL Number:MFCD01863172