Order Entry
France
ContactUsLinkComponent
2-(4-Chlorophenylthio)nicotinic acid
2-(4-Chlorophenylthio)nicotinic acid
Catalog # APOSOR21702-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Chlorophenylthio)nicotinic acid
Catalog # APOSOR21702-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:219…221 °C
Storage Temperature:Ambient
CAS Number:955-54-4
Formula:C₁₂H₈ClNO₂S
MDL Number:MFCD00052117