7-Methoxy-1-tetralone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6836-19-7
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00001696
:176.22 g/mol
Melting Pt:59…64 °C
Storage Temperature:Ambient
EINECS:229-916-0
Frequently Bought Together